Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | coiled-coil domain containing 66 | CCDC66 | -47.5kb | Anti-sense | Chr. 3 - 56557125 - 56621837 on NCBI Build GRCh38 | |
Downstream | Rho guanine nucleotide exchange factor 3 | ARHGEF3 |
GEF3, STA3, XPLN |
57.4kb | Sense | Chr. 3 - 56727418 - 57079308 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |