Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | solute carrier family 35 member E1 | SLC35E1 | -38.0kb | Sense | Chr. 19 - 16549837 - 16572382 on NCBI Build GRCh38 | |
Downstream | small integral membrane protein 7 | SMIM7 | C19orf42 | 18.5kb | Sense | Chr. 19 - 16630741 - 16660177 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |