Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | all-trans retinoic acid induced differentiation factor | ATRAID |
APR--3, APR-3, APR3, C2orf28, HSPC013, PRO240, p18 |
-26.6kb | Sense | Chr. 2 - 27212031 - 27217178 on NCBI Build GRCh38 |
Downstream | solute carrier family 30 member 3 | SLC30A3 | ZNT3 | 8.6kb | Anti-sense | Chr. 2 - 27254572 - 27275863 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |