Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | signal transducing adaptor molecule | STAM |
STAM-1, STAM1 |
-10.6kb | Anti-sense | Chr. 10 - 17644125 - 17716824 on NCBI Build GRCh38 |
Downstream | transmembrane protein 236 | TMEM236 |
FAM23A, FAM23B, bA162I21.2, bA16O1.2 |
24.3kb | Anti-sense | Chr. 10 - 17752110 - 17800916 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |