Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: P2RX4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001256796.1 | NP_001243725.1 | 3 | 568 | 106 | |
NM_001261397.1 | NP_001248326.1 | 2 | 520 | 106 | ||
NM_001261398.1 | NP_001248327.1 | 2 | 520 | 106 | ||
NM_002560.2 | NP_002551.2 | 2 | 520 | 106 | ||
NR_046372.1 | 2 | 520 | 106 | |||
GenBank mRNA | AF000234.1 | - | 2 | 116 | 106 | |
AK225604.1 | - | 2 | 216 | 106 | ||
AK293599.1 | - | 2 | 252 | 106 | ||
AK293660.1 | - | 2 | 252 | 106 | ||
AK301080.1 | - | 2 | 204 | 106 | ||
AK308549.1 | - | 2 | 252 | 106 | ||
AK308868.1 | - | 2 | 252 | 106 | ||
BC017458.1 | - | 2 | 249 | 106 | ||
BC033826.1 | - | 2 | 240 | 5216449 | 106 | |
BT019738.1 | - | 2 | 216 | 106 | ||
BT019739.1 | - | 2 | 216 | 106 | ||
BX402887.2 | - | 3 | 340 | 106 | ||
U83993.1 | - | 2 | 525 | 106 | ||
Y07684.1 | - | 2 | 243 | 106 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Ion Channel Mammalian Gene Collection Detect Genomic DNA Homo sapiens 1700 Inventoried Both primers and probe map within a single exon |