Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR03018248.hg.1 |
TC03001027.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03018248.hg.1 |
ENST00000320741.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03018248.hg.1 |
ENST00000412955.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03018248.hg.1 |
ENST00000444204.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03018248.hg.1 |
NM_001177800.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03018248.hg.1 |
NM_004797.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03018253.hg.1 |
TC03001027.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03018253.hg.1 |
ENST00000320741.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03018253.hg.1 |
ENST00000412955.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03018253.hg.1 |
ENST00000444204.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03018253.hg.1 |
NM_001177800.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03018253.hg.1 |
NM_004797.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009787.hg.1 |
ENST00000320741.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009787.hg.1 |
ENST00000320741.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009787.hg.1 |
ENST00000320741.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009787.hg.1 |
ENST00000320741.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009787.hg.1 |
ENST00000320741.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009787.hg.1 |
ENST00000320741.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009787.hg.1 |
ENST00000320741.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009787.hg.1 |
ENST00000444204.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009787.hg.1 |
ENST00000444204.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009787.hg.1 |
ENST00000444204.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009787.hg.1 |
ENST00000444204.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009787.hg.1 |
ENST00000444204.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009787.hg.1 |
ENST00000444204.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009787.hg.1 |
ENST00000444204.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009787.hg.1 |
ENST00000444204.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009787.hg.1 |
NM_001177800.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009787.hg.1 |
NM_001177800.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009787.hg.1 |
NM_001177800.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009787.hg.1 |
NM_001177800.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009787.hg.1 |
NM_001177800.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009787.hg.1 |
NM_001177800.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009787.hg.1 |
NM_001177800.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009787.hg.1 |
NM_001177800.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009787.hg.1 |
NM_004797.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009787.hg.1 |
NM_004797.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009787.hg.1 |
NM_004797.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009787.hg.1 |
NM_004797.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009787.hg.1 |
NM_004797.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009787.hg.1 |
NM_004797.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009787.hg.1 |
NM_004797.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300171264.hg.1 |
ENST00000320741.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300171264.hg.1 |
ENST00000444204.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300171264.hg.1 |
NM_001177800.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300171264.hg.1 |
NM_004797.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300171264.hg.1 |
ENST00000320741.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300171264.hg.1 |
ENST00000444204.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300171264.hg.1 |
NM_001177800.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300171264.hg.1 |
NM_004797.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300171264.hg.1 |
TC0300009787.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0300171260.hg.1 |
ENST00000320741.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300171260.hg.1 |
ENST00000444204.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300171260.hg.1 |
NM_001177800.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300171260.hg.1 |
NM_004797.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300171260.hg.1 |
ENST00000320741.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300171260.hg.1 |
ENST00000444204.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300171260.hg.1 |
NM_001177800.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300171260.hg.1 |
NM_004797.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300171260.hg.1 |
TC0300009787.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0300009787.hg.1 |
ENST00000320741.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009787.hg.1 |
ENST00000320741.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009787.hg.1 |
ENST00000320741.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009787.hg.1 |
ENST00000320741.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009787.hg.1 |
ENST00000320741.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009787.hg.1 |
ENST00000320741.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009787.hg.1 |
ENST00000320741.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009787.hg.1 |
ENST00000444204.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009787.hg.1 |
ENST00000444204.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009787.hg.1 |
ENST00000444204.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009787.hg.1 |
ENST00000444204.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009787.hg.1 |
ENST00000444204.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009787.hg.1 |
ENST00000444204.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009787.hg.1 |
ENST00000444204.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009787.hg.1 |
ENST00000444204.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009787.hg.1 |
NM_001177800.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009787.hg.1 |
NM_001177800.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009787.hg.1 |
NM_001177800.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009787.hg.1 |
NM_001177800.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009787.hg.1 |
NM_001177800.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009787.hg.1 |
NM_001177800.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009787.hg.1 |
NM_001177800.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009787.hg.1 |
NM_001177800.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009787.hg.1 |
NM_004797.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009787.hg.1 |
NM_004797.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009787.hg.1 |
NM_004797.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009787.hg.1 |
NM_004797.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009787.hg.1 |
NM_004797.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009787.hg.1 |
NM_004797.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009787.hg.1 |
NM_004797.3 |
Refseq |