Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR12010084.hg.1 |
ENST00000266744.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12010084.hg.1 |
NM_004316.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12010084.hg.1 |
TC12000796.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR12010087.hg.1 |
ENST00000266744.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12010087.hg.1 |
NM_004316.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12010087.hg.1 |
TC12000796.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR12010082.hg.1 |
ENST00000266744.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12010082.hg.1 |
NM_004316.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12010082.hg.1 |
TC12000796.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR12010085.hg.1 |
ENST00000266744.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12010085.hg.1 |
NM_004316.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12010085.hg.1 |
TC12000796.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR12010086.hg.1 |
ENST00000266744.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12010086.hg.1 |
NM_004316.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12010086.hg.1 |
TC12000796.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008650.hg.1 |
ENST00000266744.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008650.hg.1 |
ENST00000266744.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008650.hg.1 |
ENST00000266744.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008650.hg.1 |
ENST00000266744.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008650.hg.1 |
ENST00000266744.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008650.hg.1 |
NM_004316.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008650.hg.1 |
NM_004316.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008650.hg.1 |
NM_004316.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008650.hg.1 |
NM_004316.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008650.hg.1 |
NM_004316.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200163698.hg.1 |
ENST00000266744.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200163698.hg.1 |
NM_004316.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200163698.hg.1 |
ENST00000266744.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200163698.hg.1 |
NM_004316.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200163698.hg.1 |
TC1200008650.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1200163695.hg.1 |
ENST00000266744.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200163695.hg.1 |
NM_004316.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200163695.hg.1 |
ENST00000266744.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200163695.hg.1 |
NM_004316.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200163695.hg.1 |
TC1200008650.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1200163696.hg.1 |
ENST00000266744.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200163696.hg.1 |
NM_004316.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200163696.hg.1 |
ENST00000266744.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200163696.hg.1 |
NM_004316.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200163696.hg.1 |
TC1200008650.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1200008650.hg.1 |
ENST00000266744.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008650.hg.1 |
ENST00000266744.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008650.hg.1 |
ENST00000266744.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008650.hg.1 |
ENST00000266744.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008650.hg.1 |
ENST00000266744.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008650.hg.1 |
NM_004316.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008650.hg.1 |
NM_004316.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008650.hg.1 |
NM_004316.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008650.hg.1 |
NM_004316.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008650.hg.1 |
NM_004316.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200163697.hg.1 |
ENST00000266744.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200163697.hg.1 |
NM_004316.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200163697.hg.1 |
ENST00000266744.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200163697.hg.1 |
NM_004316.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200163697.hg.1 |
TC1200008650.hg.1 |
Clariom D TC |