Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012716.hg.1 |
ENST00000229379.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012716.hg.1 |
ENST00000229379.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012716.hg.1 |
ENST00000229379.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012716.hg.1 |
ENST00000229379.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012716.hg.1 |
ENST00000549525.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012716.hg.1 |
ENST00000549525.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012716.hg.1 |
ENST00000549525.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012716.hg.1 |
ENST00000550009.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012716.hg.1 |
ENST00000550009.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012716.hg.1 |
ENST00000550009.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012716.hg.1 |
NM_004373.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012716.hg.1 |
NM_004373.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012716.hg.1 |
NM_004373.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012716.hg.1 |
NM_004373.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008975.hg.1 |
ENST00000418216.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009302.hg.1 |
ENST00000404167.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12012219.hg.1 |
TC12003257.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR12012219.hg.1 |
ENST00000229379.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12012219.hg.1 |
NM_004373.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007857.hg.1 |
ENST00000620597.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008838.hg.1 |
ENST00000447525.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12012217.hg.1 |
TC12003257.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR12012217.hg.1 |
ENST00000229379.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12012217.hg.1 |
NM_004373.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12012218.hg.1 |
TC12003257.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR12012218.hg.1 |
ENST00000229379.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12012218.hg.1 |
NM_004373.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200201764.hg.1 |
ENST00000229379.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200201764.hg.1 |
ENST00000550009.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200201764.hg.1 |
NM_004373.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200201764.hg.1 |
ENST00000229379.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200201764.hg.1 |
ENST00000550009.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200201764.hg.1 |
NM_004373.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200201764.hg.1 |
TC1200012716.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1200012716.hg.1 |
ENST00000229379.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012716.hg.1 |
ENST00000229379.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012716.hg.1 |
ENST00000229379.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012716.hg.1 |
ENST00000229379.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012716.hg.1 |
ENST00000549525.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012716.hg.1 |
ENST00000549525.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012716.hg.1 |
ENST00000549525.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012716.hg.1 |
ENST00000550009.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012716.hg.1 |
ENST00000550009.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012716.hg.1 |
ENST00000550009.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012716.hg.1 |
NM_004373.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012716.hg.1 |
NM_004373.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012716.hg.1 |
NM_004373.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012716.hg.1 |
NM_004373.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200201763.hg.1 |
ENST00000229379.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200201763.hg.1 |
ENST00000550009.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200201763.hg.1 |
NM_004373.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200201763.hg.1 |
ENST00000229379.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200201763.hg.1 |
ENST00000550009.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200201763.hg.1 |
NM_004373.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200201763.hg.1 |
TC1200012716.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100008838.hg.1 |
ENST00000447525.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009302.hg.1 |
ENST00000404167.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007857.hg.1 |
ENST00000620597.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008975.hg.1 |
ENST00000418216.1 |
Ensembl |