Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012639.hg.1 |
ENST00000243108.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012639.hg.1 |
ENST00000243108.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012639.hg.1 |
ENST00000243108.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012639.hg.1 |
ENST00000243108.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012639.hg.1 |
ENST00000243108.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012639.hg.1 |
ENST00000243108.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012639.hg.1 |
ENST00000243108.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012639.hg.1 |
ENST00000243108.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012639.hg.1 |
ENST00000243108.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012639.hg.1 |
ENST00000243108.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012639.hg.1 |
ENST00000303406.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012639.hg.1 |
ENST00000303406.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012639.hg.1 |
ENST00000303406.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012639.hg.1 |
ENST00000303406.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012639.hg.1 |
ENST00000303406.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012639.hg.1 |
ENST00000303406.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012639.hg.1 |
ENST00000312492.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012639.hg.1 |
ENST00000312492.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012639.hg.1 |
ENST00000312492.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012639.hg.1 |
ENST00000312492.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012639.hg.1 |
ENST00000312492.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012639.hg.1 |
ENST00000394331.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012639.hg.1 |
ENST00000394331.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012639.hg.1 |
ENST00000394331.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012639.hg.1 |
ENST00000394331.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012639.hg.1 |
ENST00000394331.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012639.hg.1 |
ENST00000394331.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012639.hg.1 |
ENST00000394331.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012639.hg.1 |
ENST00000394331.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012639.hg.1 |
ENST00000394331.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012639.hg.1 |
ENST00000430889.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012639.hg.1 |
ENST00000430889.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012639.hg.1 |
ENST00000430889.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012639.hg.1 |
ENST00000430889.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012639.hg.1 |
ENST00000430889.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012639.hg.1 |
ENST00000507650.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012639.hg.1 |
ENST00000507650.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012639.hg.1 |
ENST00000507650.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012639.hg.1 |
ENST00000507650.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012639.hg.1 |
ENST00000512206.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012639.hg.1 |
ENST00000512206.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012639.hg.1 |
ENST00000512206.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012639.hg.1 |
NM_004503.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012639.hg.1 |
NM_004503.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012639.hg.1 |
NM_004503.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012639.hg.1 |
NM_004503.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012639.hg.1 |
NM_004503.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012639.hg.1 |
NM_004503.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012639.hg.1 |
NM_004503.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012639.hg.1 |
NM_004503.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012639.hg.1 |
NM_004503.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012639.hg.1 |
NM_014620.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012639.hg.1 |
NM_014620.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012639.hg.1 |
NM_014620.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012639.hg.1 |
NM_014620.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012639.hg.1 |
NM_014620.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012639.hg.1 |
NM_014620.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012639.hg.1 |
NM_014620.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012639.hg.1 |
NM_014620.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012639.hg.1 |
NM_018953.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012639.hg.1 |
NM_018953.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012639.hg.1 |
NM_018953.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012639.hg.1 |
NM_018953.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012639.hg.1 |
NM_153633.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012639.hg.1 |
NM_153633.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012639.hg.1 |
NM_153633.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012639.hg.1 |
NM_153633.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012639.hg.1 |
NM_153633.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012639.hg.1 |
NM_153693.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012639.hg.1 |
NM_153693.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012639.hg.1 |
NM_153693.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012639.hg.1 |
NM_153693.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012639.hg.1 |
NM_153693.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012639.hg.1 |
NM_153693.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012639.hg.1 |
NM_153693.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012639.hg.1 |
NM_153693.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012639.hg.1 |
NM_153693.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012639.hg.1 |
NR_003084.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012639.hg.1 |
NR_003084.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012639.hg.1 |
NR_003084.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012639.hg.1 |
NR_003084.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012639.hg.1 |
NR_003084.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12005903.hg.1 |
TC12003295.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR12005903.hg.1 |
ENST00000430889.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12005903.hg.1 |
NM_153633.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012639.hg.1 |
ENST00000243108.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012639.hg.1 |
ENST00000243108.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012639.hg.1 |
ENST00000243108.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012639.hg.1 |
ENST00000243108.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012639.hg.1 |
ENST00000243108.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012639.hg.1 |
ENST00000243108.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012639.hg.1 |
ENST00000243108.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012639.hg.1 |
ENST00000243108.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012639.hg.1 |
ENST00000243108.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012639.hg.1 |
ENST00000243108.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012639.hg.1 |
ENST00000303406.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012639.hg.1 |
ENST00000303406.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012639.hg.1 |
ENST00000303406.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012639.hg.1 |
ENST00000303406.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012639.hg.1 |
ENST00000303406.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012639.hg.1 |
ENST00000303406.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012639.hg.1 |
ENST00000312492.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012639.hg.1 |
ENST00000312492.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012639.hg.1 |
ENST00000312492.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012639.hg.1 |
ENST00000312492.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012639.hg.1 |
ENST00000312492.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012639.hg.1 |
ENST00000394331.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012639.hg.1 |
ENST00000394331.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012639.hg.1 |
ENST00000394331.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012639.hg.1 |
ENST00000394331.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012639.hg.1 |
ENST00000394331.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012639.hg.1 |
ENST00000394331.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012639.hg.1 |
ENST00000394331.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012639.hg.1 |
ENST00000394331.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012639.hg.1 |
ENST00000394331.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012639.hg.1 |
ENST00000430889.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012639.hg.1 |
ENST00000430889.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012639.hg.1 |
ENST00000430889.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012639.hg.1 |
ENST00000430889.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012639.hg.1 |
ENST00000430889.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012639.hg.1 |
ENST00000507650.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012639.hg.1 |
ENST00000507650.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012639.hg.1 |
ENST00000507650.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012639.hg.1 |
ENST00000507650.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012639.hg.1 |
ENST00000512206.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012639.hg.1 |
ENST00000512206.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012639.hg.1 |
ENST00000512206.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012639.hg.1 |
NM_004503.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012639.hg.1 |
NM_004503.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012639.hg.1 |
NM_004503.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012639.hg.1 |
NM_004503.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012639.hg.1 |
NM_004503.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012639.hg.1 |
NM_004503.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012639.hg.1 |
NM_004503.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012639.hg.1 |
NM_004503.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012639.hg.1 |
NM_004503.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012639.hg.1 |
NM_014620.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012639.hg.1 |
NM_014620.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012639.hg.1 |
NM_014620.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012639.hg.1 |
NM_014620.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012639.hg.1 |
NM_014620.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012639.hg.1 |
NM_014620.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012639.hg.1 |
NM_014620.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012639.hg.1 |
NM_014620.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012639.hg.1 |
NM_018953.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012639.hg.1 |
NM_018953.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012639.hg.1 |
NM_018953.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012639.hg.1 |
NM_018953.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012639.hg.1 |
NM_153633.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012639.hg.1 |
NM_153633.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012639.hg.1 |
NM_153633.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012639.hg.1 |
NM_153633.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012639.hg.1 |
NM_153633.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012639.hg.1 |
NM_153693.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012639.hg.1 |
NM_153693.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012639.hg.1 |
NM_153693.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012639.hg.1 |
NM_153693.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012639.hg.1 |
NM_153693.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012639.hg.1 |
NM_153693.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012639.hg.1 |
NM_153693.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012639.hg.1 |
NM_153693.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012639.hg.1 |
NM_153693.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012639.hg.1 |
NR_003084.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012639.hg.1 |
NR_003084.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012639.hg.1 |
NR_003084.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012639.hg.1 |
NR_003084.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012639.hg.1 |
NR_003084.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200200245.hg.1 |
ENST00000430889.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200200245.hg.1 |
NM_153633.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200200245.hg.1 |
ENST00000430889.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200200245.hg.1 |
NM_153633.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200200245.hg.1 |
TC1200012639.hg.1 |
Clariom D TC |