Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MGST2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001204366.1 | NP_001191295.1 | 4 - 5 | 567 | 99 | |
NM_001204367.1 | NP_001191296.1 | 3 - 4 | 467 | 99 | ||
NM_001204368.1 | NP_001191297.1 | 3 - 4 | 496 | 99 | ||
NM_002413.4 | NP_002404.1 | 4 - 5 | 567 | 99 | ||
XM_006714221.2 | XP_006714284.1 | 3 - 4 | 254 | 99 | ||
XM_011531972.2 | XP_011530274.1 | 3 - 4 | 233 | 99 | ||
XM_017008211.1 | XP_016863700.1 | 4 - 5 | 579 | 99 | ||
XM_017008212.1 | XP_016863701.1 | 4 - 5 | 579 | 99 | ||
XM_017008213.1 | XP_016863702.1 | 4 - 5 | 579 | 99 | ||
XM_017008214.1 | XP_016863703.1 | 3 - 4 | 254 | 99 | ||
GenBank mRNA | AA122237.1 | - | 4 - 5 | 566 | 98 | |
AK130948.1 | - | 4 - 5 | 516 | 99 | ||
AK307231.1 | - | 3 - 4 | 302 | 99 | ||
BC025416.1 | - | 4 - 5 | 481 | 3937750 | 99 | |
BE312976.1 | - | 3 - 4 | 460 | 99 | ||
CR407640.1 | - | 4 - 5 | 315 | 99 | ||
DB257575.1 | - | 4 - 5 | 331 | 99 | ||
U77604.1 | - | 4 - 5 | 326 | 99 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons |