Array Name |
Probe IDs |
Targets |
Target Type |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P27167644 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P27167635 |
- |
- |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
591441 |
- |
- |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
705099 |
- |
- |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
547796 |
- |
- |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
591082 |
- |
- |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
108458 |
- |
- |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
358348 |
- |
- |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
269197 |
- |
- |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
493259 |
- |
- |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
135455 |
AK000429.1 |
genbank_mrna_human |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
135455 |
AK096964.1 |
genbank_mrna_human |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
135455 |
AY358088.1 |
genbank_mrna_human |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
135455 |
BC005210.1 |
genbank_mrna_human |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
135455 |
BC016478.2 |
genbank_mrna_human |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
135455 |
CR595363.1 |
genbank_mrna_human |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
135455 |
CR597925.1 |
genbank_mrna_human |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
135455 |
CR600373.1 |
genbank_mrna_human |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
135455 |
CR602314.1 |
genbank_mrna_human |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
135455 |
CR602833.1 |
genbank_mrna_human |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
135455 |
CR610030.1 |
genbank_mrna_human |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
135455 |
CR610922.1 |
genbank_mrna_human |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
135455 |
CR614146.1 |
genbank_mrna_human |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
135455 |
CR615333.1 |
genbank_mrna_human |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
135455 |
CR620031.1 |
genbank_mrna_human |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
135455 |
CR623611.1 |
genbank_mrna_human |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
135455 |
CR624962.1 |
genbank_mrna_human |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
135455 |
NM_017814.1 |
refseq_mrna_human_NM |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
16427 |
- |
- |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
147234 |
- |
- |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
136856 |
- |
- |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
939602 |
- |
- |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
672134 |
- |
- |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
337820 |
- |
- |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
284208 |
- |
- |
Agilent SurePrint G3 Human CNV Microarray 2x400K |
191775 |
- |
- |
Agilent SurePrint G3 Human CNV Microarray 2x400K |
35030 |
- |
- |
Agilent SurePrint G3 Human CNV Microarray 2x400K |
292944 |
- |
- |
Agilent SurePrint G3 Human CNV Microarray 2x400K |
54623 |
- |
- |
Agilent SurePrint G3 Human CNV Microarray 2x400K |
214891 |
- |
- |
Agilent SurePrint G3 Human CNV Microarray 2x400K |
174682 |
XM_929754.2 |
refseq_mrna_human_XM |
Agilent SurePrint G3 Human CNV Microarray 2x400K |
174682 |
XM_941618.2 |
refseq_mrna_human_XM |
Agilent SurePrint G3 Human CNV Microarray 2x400K |
294955 |
- |
- |
Agilent SurePrint G3 Human CNV Microarray 2x400K |
329994 |
- |
- |
Agilent SurePrint G3 Human CNV Microarray 2x400K |
377138 |
- |
- |
Agilent SurePrint G3 Human CNV Microarray 2x400K |
59140 |
- |
- |
Agilent SurePrint G3 Human CNV Microarray 2x400K |
366949 |
- |
- |
Agilent SurePrint G3 Human CNV Microarray 2x400K |
378855 |
- |
- |
Agilent SurePrint G3 Human CNV Microarray 2x400K |
383101 |
- |
- |
Agilent Whole Human Genome Microarray 4x44K v2 |
A_33_P3330826 |
- |
- |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray |
32777 |
- |
- |
Agilent Whole Human Genome Microarray 4x44K v2 |
A_33_P3330826 |
- |
- |
SurePrint G3 Human GE Microarray G4851A |
A_33_P3330826 |
- |
- |
Agilent Human Genome 244K CGH Microarray (Alpha Test) |
A_16_P02134964 |
- |
- |
Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 |
19026 |
- |
- |
Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 |
124003 |
- |
- |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_18_P16923518 |
- |
- |