Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SYT7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_004200.3 | NP_004191.2 | 3 - 4 | 263 | 102 | |
XM_011545343.2 | XP_011543645.1 | 3 - 4 | 249 | 102 | ||
GenBank mRNA | AF038535.1 | - | 3 - 4 | 256 | 102 | |
BC125170.1 | - | 3 - 4 | 221 | 40121712 | 102 | |
BE644642.1 | - | 3 - 4 | 263 | 102 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons |