Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIM41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_033549.4 | NP_291027.3 | 3 - 4 | 1878 | 90 | |
NM_201627.2 | NP_963921.1 | 3 - 4 | 1878 | 90 | ||
XM_006714929.2 | XP_006714992.1 | 3 - 4 | 1876 | 90 | ||
XM_006714930.2 | XP_006714993.1 | 3 - 4 | 1876 | 90 | ||
XM_006714931.3 | XP_006714994.1 | 3 - 4 | 1876 | 90 | ||
XM_006714932.3 | XP_006714995.1 | 3 - 4 | 1876 | 90 | ||
GenBank mRNA | AB063180.1 | - | 3 - 4 | 904 | 90 | |
AB100366.1 | - | 3 - 4 | 1141 | 90 | ||
AB100367.1 | - | 3 - 4 | 1141 | 90 | ||
AK027601.1 | - | 3 - 4 | 1851 | 90 | ||
AX775789.1 | - | 3 - 4 | 617 | 90 | ||
AX775791.1 | - | 3 - 4 | 617 | 90 | ||
AX775793.1 | - | 3 - 4 | 1851 | 90 | ||
AX775797.1 | - | 3 - 4 | 909 | 90 | ||
AX775799.1 | - | 3 - 4 | 1851 | 90 | ||
BC018765.1 | - | 3 - 4 | 909 | 90 | ||
BC090953.1 | - | 3 - 4 | 1421 | 6017628 | 90 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons |