Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DTHD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001136536.4 | NP_001130008.2 | 7 - 8 | 1867 | 115 | |
NM_001170700.2 | NP_001164171.1 | 7 - 8 | 2022 | 115 | ||
XM_006714014.3 | XP_006714077.1 | 8 - 9 | 2501 | 115 | ||
XM_011513693.2 | XP_011511995.1 | 9 - 10 | 2529 | 115 | ||
XM_011513694.2 | XP_011511996.1 | 9 - 10 | 2466 | 115 | ||
XM_011513695.2 | XP_011511997.1 | 7 - 8 | 2339 | 115 | ||
XM_011513696.2 | XP_011511998.1 | 8 - 9 | 1912 | 115 | ||
GenBank mRNA | AK304357.1 | - | 7 - 8 | 1853 | 115 | |
AK316523.1 | - | 7 - 8 | 1853 | 115 | ||
BC157885.1 | - | 7 - 8 | 2022 | 9057096 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons |