Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DTHD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001136536.4 | NP_001130008.2 | 5 - 6 | 1332 | 119 | |
NM_001170700.2 | NP_001164171.1 | 5 - 6 | 1487 | 119 | ||
XM_006714014.3 | XP_006714077.1 | 6 - 7 | 1966 | 119 | ||
XM_011513693.2 | XP_011511995.1 | 7 - 8 | 1994 | 119 | ||
XM_011513694.2 | XP_011511996.1 | 7 - 8 | 1931 | 119 | ||
XM_011513696.2 | XP_011511998.1 | 6 - 7 | 1377 | 119 | ||
XM_017008191.1 | XP_016863680.1 | 7 - 8 | 1994 | 119 | ||
XM_017008192.1 | XP_016863681.1 | 7 - 8 | 1996 | 119 | ||
GenBank mRNA | AK131492.1 | - | 5 - 6 | 1325 | 119 | |
AK304357.1 | - | 5 - 6 | 1318 | 119 | ||
AK316523.1 | - | 5 - 6 | 1318 | 119 | ||
BC157885.1 | - | 5 - 6 | 1487 | 9057096 | 119 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons |