Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR16006297.hg.1 |
ENST00000320990.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16006297.hg.1 |
ENST00000394636.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16006297.hg.1 |
ENST00000558597.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16006297.hg.1 |
NM_001252197.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16006297.hg.1 |
NM_005853.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16006297.hg.1 |
TC16000451.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007918.hg.1 |
ENST00000320990.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007918.hg.1 |
ENST00000320990.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007918.hg.1 |
ENST00000320990.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007918.hg.1 |
ENST00000320990.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007918.hg.1 |
ENST00000320990.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007918.hg.1 |
ENST00000320990.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007918.hg.1 |
ENST00000320990.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007918.hg.1 |
ENST00000394636.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007918.hg.1 |
ENST00000394636.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007918.hg.1 |
ENST00000394636.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007918.hg.1 |
ENST00000394636.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007918.hg.1 |
ENST00000394636.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007918.hg.1 |
ENST00000394636.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007918.hg.1 |
ENST00000394636.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007918.hg.1 |
ENST00000394636.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007918.hg.1 |
ENST00000558597.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007918.hg.1 |
ENST00000558597.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007918.hg.1 |
ENST00000558597.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007918.hg.1 |
ENST00000558597.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007918.hg.1 |
ENST00000558597.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007918.hg.1 |
ENST00000558597.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007918.hg.1 |
ENST00000558597.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007918.hg.1 |
ENST00000560154.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007918.hg.1 |
ENST00000560154.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007918.hg.1 |
ENST00000560154.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007918.hg.1 |
ENST00000560154.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007918.hg.1 |
ENST00000620085.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007918.hg.1 |
ENST00000620085.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007918.hg.1 |
ENST00000620085.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007918.hg.1 |
ENST00000620085.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007918.hg.1 |
NM_001252197.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007918.hg.1 |
NM_001252197.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007918.hg.1 |
NM_001252197.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007918.hg.1 |
NM_001252197.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007918.hg.1 |
NM_001252197.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007918.hg.1 |
NM_001252197.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007918.hg.1 |
NM_001252197.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007918.hg.1 |
NM_005853.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007918.hg.1 |
NM_005853.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007918.hg.1 |
NM_005853.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007918.hg.1 |
NM_005853.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007918.hg.1 |
NM_005853.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007918.hg.1 |
NM_005853.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007918.hg.1 |
NM_005853.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16006298.hg.1 |
ENST00000320990.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16006298.hg.1 |
ENST00000394636.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16006298.hg.1 |
NM_001252197.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16006298.hg.1 |
NM_005853.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16006298.hg.1 |
TC16000451.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR16006296.hg.1 |
ENST00000320990.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16006296.hg.1 |
ENST00000394636.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16006296.hg.1 |
ENST00000447390.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16006296.hg.1 |
ENST00000558597.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16006296.hg.1 |
ENST00000560154.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16006296.hg.1 |
NM_001252197.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16006296.hg.1 |
NM_005853.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16006296.hg.1 |
TC16000451.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1600157653.hg.1 |
ENST00000320990.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600157653.hg.1 |
ENST00000394636.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600157653.hg.1 |
ENST00000558597.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600157653.hg.1 |
ENST00000560154.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600157653.hg.1 |
ENST00000620085.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600157653.hg.1 |
NM_001252197.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600157653.hg.1 |
NM_005853.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600157653.hg.1 |
ENST00000320990.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600157653.hg.1 |
ENST00000394636.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600157653.hg.1 |
ENST00000558597.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600157653.hg.1 |
ENST00000560154.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600157653.hg.1 |
ENST00000620085.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600157653.hg.1 |
NM_001252197.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600157653.hg.1 |
NM_005853.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600157653.hg.1 |
TC1600007918.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1600157655.hg.1 |
ENST00000320990.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600157655.hg.1 |
ENST00000394636.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600157655.hg.1 |
NM_001252197.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600157655.hg.1 |
NM_005853.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600157655.hg.1 |
ENST00000320990.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600157655.hg.1 |
ENST00000394636.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600157655.hg.1 |
NM_001252197.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600157655.hg.1 |
NM_005853.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600157655.hg.1 |
TC1600007918.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1600157652.hg.1 |
ENST00000320990.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600157652.hg.1 |
ENST00000394636.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600157652.hg.1 |
ENST00000558597.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600157652.hg.1 |
ENST00000560154.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600157652.hg.1 |
ENST00000620085.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600157652.hg.1 |
NM_001252197.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600157652.hg.1 |
NM_005853.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600157652.hg.1 |
ENST00000320990.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600157652.hg.1 |
ENST00000394636.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600157652.hg.1 |
ENST00000558597.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600157652.hg.1 |
ENST00000560154.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600157652.hg.1 |
ENST00000620085.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600157652.hg.1 |
NM_001252197.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600157652.hg.1 |
NM_005853.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600157652.hg.1 |
TC1600007918.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1600157654.hg.1 |
ENST00000320990.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600157654.hg.1 |
ENST00000394636.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600157654.hg.1 |
ENST00000558597.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600157654.hg.1 |
NM_001252197.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600157654.hg.1 |
NM_005853.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600157654.hg.1 |
ENST00000320990.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600157654.hg.1 |
ENST00000394636.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600157654.hg.1 |
ENST00000558597.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600157654.hg.1 |
NM_001252197.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600157654.hg.1 |
NM_005853.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600157654.hg.1 |
TC1600007918.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1600007918.hg.1 |
ENST00000320990.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007918.hg.1 |
ENST00000320990.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007918.hg.1 |
ENST00000320990.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007918.hg.1 |
ENST00000320990.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007918.hg.1 |
ENST00000320990.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007918.hg.1 |
ENST00000320990.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007918.hg.1 |
ENST00000320990.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007918.hg.1 |
ENST00000394636.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007918.hg.1 |
ENST00000394636.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007918.hg.1 |
ENST00000394636.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007918.hg.1 |
ENST00000394636.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007918.hg.1 |
ENST00000394636.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007918.hg.1 |
ENST00000394636.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007918.hg.1 |
ENST00000394636.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007918.hg.1 |
ENST00000394636.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007918.hg.1 |
ENST00000558597.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007918.hg.1 |
ENST00000558597.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007918.hg.1 |
ENST00000558597.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007918.hg.1 |
ENST00000558597.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007918.hg.1 |
ENST00000558597.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007918.hg.1 |
ENST00000558597.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007918.hg.1 |
ENST00000558597.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007918.hg.1 |
ENST00000560154.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007918.hg.1 |
ENST00000560154.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007918.hg.1 |
ENST00000560154.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007918.hg.1 |
ENST00000560154.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007918.hg.1 |
ENST00000620085.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007918.hg.1 |
ENST00000620085.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007918.hg.1 |
ENST00000620085.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007918.hg.1 |
ENST00000620085.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007918.hg.1 |
NM_001252197.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007918.hg.1 |
NM_001252197.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007918.hg.1 |
NM_001252197.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007918.hg.1 |
NM_001252197.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007918.hg.1 |
NM_001252197.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007918.hg.1 |
NM_001252197.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007918.hg.1 |
NM_001252197.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007918.hg.1 |
NM_005853.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007918.hg.1 |
NM_005853.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007918.hg.1 |
NM_005853.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007918.hg.1 |
NM_005853.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007918.hg.1 |
NM_005853.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007918.hg.1 |
NM_005853.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007918.hg.1 |
NM_005853.5 |
Refseq |