Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SKAP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_017012771.1 | XP_016868260.1 | 13 | 1807 | 79 | |
GenBank mRNA | DQ218281.1 | - | 1 | 227 | 79 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | chromosome 7 open reading frame 71 | C7orf71 | -3.2kb | Anti-sense | Chr. 7 - 26635582 - 26647272 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |