Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NR2C2AP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001300945.1 | NP_001287874.1 | 5 - 6 | 784 | 70 | |
XM_011527688.2 | XP_011525990.1 | 5 - 6 | 814 | 70 | ||
GenBank mRNA | AA588235.1 | - | 1 - 2 | 70 | ||
AK301426.1 | - | 5 - 6 | 712 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Homo sapiens Probe spans exons |