Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HAS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001297436.1 | NP_001284365.1 | 4 - 5 | 1116 | 95 | |
NM_001523.3 | NP_001514.2 | 4 - 5 | 1119 | 95 | ||
GenBank mRNA | BC035837.1 | - | 4 - 5 | 1098 | 5589083 | 95 |
BX371636.2 | - | 1 - 2 | 95 | |||
D84424.1 | - | 3 - 4 | 1103 | 95 | ||
U59269.1 | - | 4 - 5 | 1095 | 95 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Homo sapiens Probe spans exons |