Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006692.hg.1 |
ENST00000320912.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006692.hg.1 |
ENST00000320912.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006692.hg.1 |
ENST00000320912.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006692.hg.1 |
ENST00000320912.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006692.hg.1 |
ENST00000320912.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006692.hg.1 |
ENST00000320912.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006692.hg.1 |
ENST00000320912.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006692.hg.1 |
ENST00000320912.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006692.hg.1 |
ENST00000382581.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006692.hg.1 |
ENST00000382581.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006692.hg.1 |
ENST00000382581.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006692.hg.1 |
ENST00000382581.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006692.hg.1 |
ENST00000382581.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006692.hg.1 |
ENST00000382581.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006692.hg.1 |
ENST00000382581.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006692.hg.1 |
ENST00000382581.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006692.hg.1 |
ENST00000382581.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006692.hg.1 |
ENST00000507420.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006692.hg.1 |
ENST00000507420.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006692.hg.1 |
ENST00000507420.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006692.hg.1 |
ENST00000507420.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006692.hg.1 |
ENST00000507420.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006692.hg.1 |
ENST00000512914.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006692.hg.1 |
ENST00000512914.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006692.hg.1 |
ENST00000512914.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006692.hg.1 |
ENST00000512914.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006692.hg.1 |
ENST00000512914.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006692.hg.1 |
ENST00000512914.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006692.hg.1 |
ENST00000512914.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006692.hg.1 |
ENST00000512914.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006692.hg.1 |
ENST00000512914.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006692.hg.1 |
ENST00000617365.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006692.hg.1 |
ENST00000617365.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006692.hg.1 |
ENST00000617365.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006692.hg.1 |
ENST00000617365.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006692.hg.1 |
ENST00000617365.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006692.hg.1 |
ENST00000617365.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006692.hg.1 |
ENST00000617365.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006692.hg.1 |
ENST00000617365.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006692.hg.1 |
ENST00000617365.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006692.hg.1 |
NM_001272053.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006692.hg.1 |
NM_001272053.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006692.hg.1 |
NM_001272053.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006692.hg.1 |
NM_001272053.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006692.hg.1 |
NM_001272053.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006692.hg.1 |
NM_001272053.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006692.hg.1 |
NM_001272053.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006692.hg.1 |
NM_001272053.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006692.hg.1 |
NM_001272054.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006692.hg.1 |
NM_001272054.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006692.hg.1 |
NM_001272054.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006692.hg.1 |
NM_001272054.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006692.hg.1 |
NM_001272054.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006692.hg.1 |
NM_001272054.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006692.hg.1 |
NM_001272054.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006692.hg.1 |
NM_001272054.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006692.hg.1 |
NM_001272054.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006692.hg.1 |
NM_033296.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006692.hg.1 |
NM_033296.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006692.hg.1 |
NM_033296.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006692.hg.1 |
NM_033296.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006692.hg.1 |
NM_033296.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006692.hg.1 |
NM_033296.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006692.hg.1 |
NM_033296.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006692.hg.1 |
NM_033296.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006692.hg.1 |
NM_033296.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006692.hg.1 |
NM_033296.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400006692.hg.1 |
ENST00000320912.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006692.hg.1 |
ENST00000320912.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006692.hg.1 |
ENST00000320912.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006692.hg.1 |
ENST00000320912.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006692.hg.1 |
ENST00000320912.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006692.hg.1 |
ENST00000320912.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006692.hg.1 |
ENST00000320912.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006692.hg.1 |
ENST00000320912.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006692.hg.1 |
ENST00000382581.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006692.hg.1 |
ENST00000382581.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006692.hg.1 |
ENST00000382581.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006692.hg.1 |
ENST00000382581.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006692.hg.1 |
ENST00000382581.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006692.hg.1 |
ENST00000382581.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006692.hg.1 |
ENST00000382581.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006692.hg.1 |
ENST00000382581.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006692.hg.1 |
ENST00000382581.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006692.hg.1 |
ENST00000507420.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006692.hg.1 |
ENST00000507420.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006692.hg.1 |
ENST00000507420.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006692.hg.1 |
ENST00000507420.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006692.hg.1 |
ENST00000507420.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006692.hg.1 |
ENST00000512914.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006692.hg.1 |
ENST00000512914.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006692.hg.1 |
ENST00000512914.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006692.hg.1 |
ENST00000512914.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006692.hg.1 |
ENST00000512914.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006692.hg.1 |
ENST00000512914.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006692.hg.1 |
ENST00000512914.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006692.hg.1 |
ENST00000512914.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006692.hg.1 |
ENST00000512914.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006692.hg.1 |
ENST00000617365.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006692.hg.1 |
ENST00000617365.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006692.hg.1 |
ENST00000617365.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006692.hg.1 |
ENST00000617365.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006692.hg.1 |
ENST00000617365.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006692.hg.1 |
ENST00000617365.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006692.hg.1 |
ENST00000617365.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006692.hg.1 |
ENST00000617365.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006692.hg.1 |
ENST00000617365.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006692.hg.1 |
NM_001272053.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400006692.hg.1 |
NM_001272053.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400006692.hg.1 |
NM_001272053.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400006692.hg.1 |
NM_001272053.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400006692.hg.1 |
NM_001272053.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400006692.hg.1 |
NM_001272053.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400006692.hg.1 |
NM_001272053.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400006692.hg.1 |
NM_001272053.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400006692.hg.1 |
NM_001272054.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400006692.hg.1 |
NM_001272054.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400006692.hg.1 |
NM_001272054.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400006692.hg.1 |
NM_001272054.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400006692.hg.1 |
NM_001272054.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400006692.hg.1 |
NM_001272054.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400006692.hg.1 |
NM_001272054.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400006692.hg.1 |
NM_001272054.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400006692.hg.1 |
NM_001272054.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400006692.hg.1 |
NM_033296.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400006692.hg.1 |
NM_033296.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400006692.hg.1 |
NM_033296.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400006692.hg.1 |
NM_033296.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400006692.hg.1 |
NM_033296.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400006692.hg.1 |
NM_033296.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400006692.hg.1 |
NM_033296.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400006692.hg.1 |
NM_033296.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400006692.hg.1 |
NM_033296.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400006692.hg.1 |
NM_033296.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400146384.hg.1 |
ENST00000382581.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400146384.hg.1 |
ENST00000507420.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400146384.hg.1 |
ENST00000512914.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400146384.hg.1 |
ENST00000617365.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400146384.hg.1 |
NM_001272054.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400146384.hg.1 |
NM_033296.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400146384.hg.1 |
ENST00000382581.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400146384.hg.1 |
ENST00000507420.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400146384.hg.1 |
ENST00000512914.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400146384.hg.1 |
ENST00000617365.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400146384.hg.1 |
NM_001272054.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400146384.hg.1 |
NM_033296.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400146384.hg.1 |
TC0400006692.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0400146386.hg.1 |
ENST00000320912.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400146386.hg.1 |
ENST00000382581.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400146386.hg.1 |
ENST00000507420.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400146386.hg.1 |
ENST00000512914.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400146386.hg.1 |
ENST00000617365.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400146386.hg.1 |
NM_001272053.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400146386.hg.1 |
NM_001272054.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400146386.hg.1 |
NM_033296.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400146386.hg.1 |
ENST00000320912.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400146386.hg.1 |
ENST00000382581.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400146386.hg.1 |
ENST00000507420.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400146386.hg.1 |
ENST00000512914.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400146386.hg.1 |
ENST00000617365.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400146386.hg.1 |
NM_001272053.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400146386.hg.1 |
NM_001272054.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400146386.hg.1 |
NM_033296.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400146386.hg.1 |
TC0400006692.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0400146387.hg.1 |
ENST00000320912.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400146387.hg.1 |
ENST00000382581.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400146387.hg.1 |
ENST00000507420.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400146387.hg.1 |
ENST00000512914.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400146387.hg.1 |
ENST00000617365.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400146387.hg.1 |
NM_001272053.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400146387.hg.1 |
NM_001272054.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400146387.hg.1 |
NM_033296.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400146387.hg.1 |
ENST00000320912.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400146387.hg.1 |
ENST00000382581.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400146387.hg.1 |
ENST00000507420.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400146387.hg.1 |
ENST00000512914.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400146387.hg.1 |
ENST00000617365.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400146387.hg.1 |
NM_001272053.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400146387.hg.1 |
NM_001272054.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400146387.hg.1 |
NM_033296.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400146387.hg.1 |
TC0400006692.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0400146385.hg.1 |
ENST00000382581.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400146385.hg.1 |
ENST00000512914.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400146385.hg.1 |
ENST00000617365.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400146385.hg.1 |
NM_001272054.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400146385.hg.1 |
NM_033296.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400146385.hg.1 |
ENST00000382581.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400146385.hg.1 |
ENST00000512914.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400146385.hg.1 |
ENST00000617365.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400146385.hg.1 |
NM_001272054.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400146385.hg.1 |
NM_033296.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400146385.hg.1 |
TC0400006692.hg.1 |
Clariom D TC |