Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPIA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001300981.1 | NP_001287910.1 | 6 | 2280 | 96 | |
NM_021130.4 | NP_066953.1 | 5 | 2123 | 96 | ||
GenBank mRNA | AK130101.1 | - | 5 | 2080 | 96 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |