Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011550.hg.1 |
ENST00000564618.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011550.hg.1 |
ENST00000564618.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011550.hg.1 |
ENST00000564618.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011550.hg.1 |
ENST00000564618.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011550.hg.1 |
ENST00000567528.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011550.hg.1 |
ENST00000567528.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011550.hg.1 |
ENST00000567528.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011550.hg.1 |
ENST00000567528.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011550.hg.1 |
ENST00000567528.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011550.hg.1 |
ENST00000567528.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011550.hg.1 |
ENST00000567528.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011550.hg.1 |
ENST00000567528.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011550.hg.1 |
ENST00000569125.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011550.hg.1 |
ENST00000569125.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011550.hg.1 |
ENST00000569125.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011550.hg.1 |
ENST00000569125.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011550.hg.1 |
ENST00000569125.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011550.hg.1 |
NR_109960.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011550.hg.1 |
NR_109960.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011550.hg.1 |
NR_109960.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011550.hg.1 |
NR_109960.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011550.hg.1 |
ENST00000564618.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011550.hg.1 |
ENST00000564618.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011550.hg.1 |
ENST00000564618.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011550.hg.1 |
ENST00000564618.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011550.hg.1 |
ENST00000567528.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011550.hg.1 |
ENST00000567528.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011550.hg.1 |
ENST00000567528.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011550.hg.1 |
ENST00000567528.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011550.hg.1 |
ENST00000567528.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011550.hg.1 |
ENST00000567528.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011550.hg.1 |
ENST00000567528.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011550.hg.1 |
ENST00000567528.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011550.hg.1 |
ENST00000569125.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011550.hg.1 |
ENST00000569125.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011550.hg.1 |
ENST00000569125.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011550.hg.1 |
ENST00000569125.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011550.hg.1 |
ENST00000569125.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011550.hg.1 |
NR_109960.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011550.hg.1 |
NR_109960.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011550.hg.1 |
NR_109960.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011550.hg.1 |
NR_109960.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600193908.hg.1 |
ENST00000564618.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600193908.hg.1 |
ENST00000567528.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600193908.hg.1 |
ENST00000567528.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600193908.hg.1 |
NR_109960.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600193908.hg.1 |
ENST00000564618.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600193908.hg.1 |
ENST00000567528.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600193908.hg.1 |
ENST00000567528.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600193908.hg.1 |
NR_109960.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600193908.hg.1 |
TC1600011550.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1600193909.hg.1 |
ENST00000564618.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600193909.hg.1 |
ENST00000567528.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600193909.hg.1 |
ENST00000567528.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600193909.hg.1 |
ENST00000569125.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600193909.hg.1 |
NR_109960.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600193909.hg.1 |
ENST00000564618.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600193909.hg.1 |
ENST00000567528.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600193909.hg.1 |
ENST00000567528.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600193909.hg.1 |
ENST00000569125.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600193909.hg.1 |
NR_109960.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600193909.hg.1 |
TC1600011550.hg.1 |
Clariom D TC |