Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SFXN4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_110305.1 | 13 - 14 | 1072 | 107 | ||
GenBank mRNA | AA401250.1 | - | 1 - 2 | 107 | ||
AY269785.1 | - | 13 - 14 | 1030 | 107 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | family with sequence similarity 45 member A | FAM45A | -2.9kb | Anti-sense | Chr. 10 - 119104065 - 119137984 on NCBI Build GRCh38 | |
Downstream | peroxiredoxin 3 | PRDX3 |
AOP-1, AOP1, HBC189, MER5, PRO1748, SP-22, prx-III |
25.3kb | Sense | Chr. 10 - 119167699 - 119178865 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Homo sapiens Probe spans exons |