Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008541.hg.1 |
ENST00000412442.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008541.hg.1 |
ENST00000442192.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008541.hg.1 |
ENST00000442192.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008541.hg.1 |
ENST00000442192.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008541.hg.1 |
ENST00000442192.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008541.hg.1 |
ENST00000442192.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008541.hg.1 |
ENST00000442192.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008541.hg.1 |
ENST00000442192.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008541.hg.1 |
ENST00000442192.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC14_GL000009v2_random00006453.hg.1 |
ENST00000412492.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC14_GL000009v2_random00006453.hg.1 |
ENST00000412492.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC14_GL000009v2_random00006453.hg.1 |
ENST00000412492.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC14_GL000009v2_random00006453.hg.1 |
ENST00000415543.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC14_GL000009v2_random00006453.hg.1 |
ENST00000415543.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC14_GL000009v2_random00006453.hg.1 |
ENST00000415543.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC14_GL000009v2_random00006453.hg.1 |
ENST00000415543.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC14_GL000009v2_random00006453.hg.1 |
ENST00000415543.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC14_GL000009v2_random00006453.hg.1 |
ENST00000415543.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC14_GL000009v2_random00006453.hg.1 |
ENST00000421640.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC14_GL000009v2_random00006453.hg.1 |
ENST00000421640.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC14_GL000009v2_random00006453.hg.1 |
ENST00000421640.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC14_GL000009v2_random00006453.hg.1 |
ENST00000421640.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC14_GL000009v2_random00006453.hg.1 |
ENST00000421640.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC14_GL000009v2_random00006453.hg.1 |
ENST00000421640.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC14_GL000009v2_random00006453.hg.1 |
ENST00000423394.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC14_GL000009v2_random00006453.hg.1 |
ENST00000423394.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC14_GL000009v2_random00006453.hg.1 |
ENST00000423394.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC14_GL000009v2_random00006453.hg.1 |
ENST00000428624.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC14_GL000009v2_random00006453.hg.1 |
ENST00000428624.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC14_GL000009v2_random00006453.hg.1 |
ENST00000428624.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC14_GL000009v2_random00006453.hg.1 |
ENST00000428624.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC14_GL000009v2_random00006453.hg.1 |
ENST00000428624.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC14_GL000009v2_random00006453.hg.1 |
ENST00000428624.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC14_GL000009v2_random00006453.hg.1 |
ENST00000428624.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC14_GL000009v2_random00006453.hg.1 |
ENST00000428624.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC14_GL000009v2_random00006453.hg.1 |
ENST00000428624.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC14_GL000009v2_random00006453.hg.1 |
ENST00000431700.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC14_GL000009v2_random00006453.hg.1 |
ENST00000431700.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC14_GL000009v2_random00006453.hg.1 |
ENST00000431700.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC14_GL000009v2_random00006453.hg.1 |
ENST00000431700.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC14_GL000009v2_random00006453.hg.1 |
ENST00000448665.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC14_GL000009v2_random00006453.hg.1 |
ENST00000448665.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC14_GL000009v2_random00006453.hg.1 |
ENST00000448665.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC14_GL000009v2_random00006453.hg.1 |
ENST00000448665.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC14_GL000009v2_random00006453.hg.1 |
ENST00000448844.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC14_GL000009v2_random00006453.hg.1 |
ENST00000448844.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC14_GL000009v2_random00006453.hg.1 |
ENST00000448844.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC14_GL000009v2_random00006453.hg.1 |
ENST00000448844.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC14_GL000009v2_random00006453.hg.1 |
ENST00000448844.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC14_GL000009v2_random00006453.hg.1 |
ENST00000448844.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC14_GL000009v2_random00006453.hg.1 |
ENST00000448844.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC14_GL000009v2_random00006453.hg.1 |
ENST00000448844.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC14_GL000009v2_random00006453.hg.1 |
ENST00000448844.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC14_GL000009v2_random00006453.hg.1 |
ENST00000448844.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC14_GL000009v2_random00006453.hg.1 |
ENST00000456460.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC14_GL000009v2_random00006453.hg.1 |
ENST00000456460.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC14_GL000009v2_random00006453.hg.1 |
ENST00000456460.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC14_GL000009v2_random00006453.hg.1 |
ENST00000456460.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC14_GL000009v2_random00006453.hg.1 |
ENST00000456460.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC14_GL000009v2_random00006453.hg.1 |
ENST00000458155.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC14_GL000009v2_random00006453.hg.1 |
ENST00000458155.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC14_GL000009v2_random00006453.hg.1 |
ENST00000458155.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC14_GL000009v2_random00006453.hg.1 |
ENST00000458155.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC14_GL000009v2_random00006453.hg.1 |
ENST00000458155.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC14_GL000009v2_random00006453.hg.1 |
ENST00000458155.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC14_GL000009v2_random00006453.hg.1 |
ENST00000458155.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC14_GL000009v2_random00006453.hg.1 |
ENST00000458155.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC14_GL000009v2_random00006453.hg.1 |
ENST00000458155.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008541.hg.1 |
ENST00000412442.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008541.hg.1 |
ENST00000442192.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008541.hg.1 |
ENST00000442192.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008541.hg.1 |
ENST00000442192.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008541.hg.1 |
ENST00000442192.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008541.hg.1 |
ENST00000442192.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008541.hg.1 |
ENST00000442192.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008541.hg.1 |
ENST00000442192.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008541.hg.1 |
ENST00000442192.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200144057.hg.1 |
TC2200006442.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2200144057.hg.1 |
NR_110761.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200144057.hg.1 |
NR_110761.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200144056.hg.1 |
TC2200006442.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2200144056.hg.1 |
NR_110761.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200144056.hg.1 |
NR_110761.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC14_GL000009v2_random00006453.hg.1 |
ENST00000412492.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC14_GL000009v2_random00006453.hg.1 |
ENST00000412492.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC14_GL000009v2_random00006453.hg.1 |
ENST00000412492.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC14_GL000009v2_random00006453.hg.1 |
ENST00000415543.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC14_GL000009v2_random00006453.hg.1 |
ENST00000415543.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC14_GL000009v2_random00006453.hg.1 |
ENST00000415543.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC14_GL000009v2_random00006453.hg.1 |
ENST00000415543.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC14_GL000009v2_random00006453.hg.1 |
ENST00000415543.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC14_GL000009v2_random00006453.hg.1 |
ENST00000415543.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC14_GL000009v2_random00006453.hg.1 |
ENST00000421640.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC14_GL000009v2_random00006453.hg.1 |
ENST00000421640.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC14_GL000009v2_random00006453.hg.1 |
ENST00000421640.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC14_GL000009v2_random00006453.hg.1 |
ENST00000421640.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC14_GL000009v2_random00006453.hg.1 |
ENST00000421640.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC14_GL000009v2_random00006453.hg.1 |
ENST00000421640.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC14_GL000009v2_random00006453.hg.1 |
ENST00000423394.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC14_GL000009v2_random00006453.hg.1 |
ENST00000423394.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC14_GL000009v2_random00006453.hg.1 |
ENST00000423394.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC14_GL000009v2_random00006453.hg.1 |
ENST00000428624.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC14_GL000009v2_random00006453.hg.1 |
ENST00000428624.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC14_GL000009v2_random00006453.hg.1 |
ENST00000428624.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC14_GL000009v2_random00006453.hg.1 |
ENST00000428624.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC14_GL000009v2_random00006453.hg.1 |
ENST00000428624.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC14_GL000009v2_random00006453.hg.1 |
ENST00000428624.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC14_GL000009v2_random00006453.hg.1 |
ENST00000428624.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC14_GL000009v2_random00006453.hg.1 |
ENST00000428624.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC14_GL000009v2_random00006453.hg.1 |
ENST00000428624.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC14_GL000009v2_random00006453.hg.1 |
ENST00000431700.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC14_GL000009v2_random00006453.hg.1 |
ENST00000431700.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC14_GL000009v2_random00006453.hg.1 |
ENST00000431700.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC14_GL000009v2_random00006453.hg.1 |
ENST00000431700.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC14_GL000009v2_random00006453.hg.1 |
ENST00000448665.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC14_GL000009v2_random00006453.hg.1 |
ENST00000448665.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC14_GL000009v2_random00006453.hg.1 |
ENST00000448665.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC14_GL000009v2_random00006453.hg.1 |
ENST00000448665.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC14_GL000009v2_random00006453.hg.1 |
ENST00000448844.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC14_GL000009v2_random00006453.hg.1 |
ENST00000448844.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC14_GL000009v2_random00006453.hg.1 |
ENST00000448844.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC14_GL000009v2_random00006453.hg.1 |
ENST00000448844.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC14_GL000009v2_random00006453.hg.1 |
ENST00000448844.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC14_GL000009v2_random00006453.hg.1 |
ENST00000448844.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC14_GL000009v2_random00006453.hg.1 |
ENST00000448844.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC14_GL000009v2_random00006453.hg.1 |
ENST00000448844.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC14_GL000009v2_random00006453.hg.1 |
ENST00000448844.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC14_GL000009v2_random00006453.hg.1 |
ENST00000448844.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC14_GL000009v2_random00006453.hg.1 |
ENST00000456460.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC14_GL000009v2_random00006453.hg.1 |
ENST00000456460.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC14_GL000009v2_random00006453.hg.1 |
ENST00000456460.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC14_GL000009v2_random00006453.hg.1 |
ENST00000456460.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC14_GL000009v2_random00006453.hg.1 |
ENST00000456460.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC14_GL000009v2_random00006453.hg.1 |
ENST00000458155.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC14_GL000009v2_random00006453.hg.1 |
ENST00000458155.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC14_GL000009v2_random00006453.hg.1 |
ENST00000458155.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC14_GL000009v2_random00006453.hg.1 |
ENST00000458155.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC14_GL000009v2_random00006453.hg.1 |
ENST00000458155.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC14_GL000009v2_random00006453.hg.1 |
ENST00000458155.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC14_GL000009v2_random00006453.hg.1 |
ENST00000458155.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC14_GL000009v2_random00006453.hg.1 |
ENST00000458155.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC14_GL000009v2_random00006453.hg.1 |
ENST00000458155.1 |
Ensembl |