Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015414.hg.1 |
ENST00000421254.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015414.hg.1 |
ENST00000421254.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015414.hg.1 |
ENST00000445523.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015414.hg.1 |
ENST00000445523.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015414.hg.1 |
ENST00000445523.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015414.hg.1 |
ENST00000445523.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015414.hg.1 |
ENST00000445523.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015414.hg.1 |
NR_110786.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015414.hg.1 |
NR_110786.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015414.hg.1 |
NR_110786.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015414.hg.1 |
NR_110786.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015414.hg.1 |
NR_110786.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100215141.hg.1 |
ENST00000445523.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100215141.hg.1 |
NR_110786.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100215141.hg.1 |
ENST00000445523.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100215141.hg.1 |
NR_110786.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100215141.hg.1 |
TC0100015414.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100215142.hg.1 |
ENST00000445523.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100215142.hg.1 |
NR_110786.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100215142.hg.1 |
ENST00000445523.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100215142.hg.1 |
NR_110786.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100215142.hg.1 |
TC0100015414.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100015414.hg.1 |
ENST00000421254.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015414.hg.1 |
ENST00000421254.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015414.hg.1 |
ENST00000445523.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015414.hg.1 |
ENST00000445523.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015414.hg.1 |
ENST00000445523.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015414.hg.1 |
ENST00000445523.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015414.hg.1 |
ENST00000445523.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015414.hg.1 |
NR_110786.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015414.hg.1 |
NR_110786.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015414.hg.1 |
NR_110786.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015414.hg.1 |
NR_110786.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015414.hg.1 |
NR_110786.1 |
Refseq |