Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009853.hg.1 |
ENST00000502156.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009853.hg.1 |
ENST00000502156.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009853.hg.1 |
ENST00000502156.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009853.hg.1 |
ENST00000502156.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009853.hg.1 |
ENST00000502156.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009853.hg.1 |
NR_120345.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009853.hg.1 |
NR_120345.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009853.hg.1 |
NR_120345.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009853.hg.1 |
NR_120345.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009853.hg.1 |
NR_120345.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500173091.hg.1 |
ENST00000502156.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500173091.hg.1 |
NR_120345.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500173091.hg.1 |
ENST00000502156.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500173091.hg.1 |
NR_120345.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500173091.hg.1 |
TC1500009853.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1500009853.hg.1 |
ENST00000502156.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009853.hg.1 |
ENST00000502156.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009853.hg.1 |
ENST00000502156.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009853.hg.1 |
ENST00000502156.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009853.hg.1 |
ENST00000502156.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009853.hg.1 |
NR_120345.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009853.hg.1 |
NR_120345.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009853.hg.1 |
NR_120345.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009853.hg.1 |
NR_120345.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009853.hg.1 |
NR_120345.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500173088.hg.1 |
ENST00000502156.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500173088.hg.1 |
NR_120345.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500173088.hg.1 |
ENST00000502156.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500173088.hg.1 |
NR_120345.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500173088.hg.1 |
TC1500009853.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1500173092.hg.1 |
ENST00000502156.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500173092.hg.1 |
NR_120345.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500173092.hg.1 |
ENST00000502156.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500173092.hg.1 |
NR_120345.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500173092.hg.1 |
TC1500009853.hg.1 |
Clariom D TC |