Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GUSB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_000181.3 | NP_000172.2 | 12 | 2176 | 95 | |
NM_001284290.1 | NP_001271219.1 | 10 | 1738 | 95 | ||
NM_001293104.1 | NP_001280033.1 | 11 | 1991 | 95 | ||
NM_001293105.1 | NP_001280034.1 | 10 | 1848 | 95 | ||
NR_120531.1 | 11 | 2091 | 95 | |||
XM_005250297.3 | XP_005250354.1 | 11 | 2018 | 95 | ||
XM_011516113.1 | XP_011514415.1 | 11 | 1920 | 95 | ||
XM_011516114.1 | XP_011514416.1 | 11 | 1848 | 95 | ||
XM_017012091.1 | XP_016867580.1 | 10 | 1767 | 95 | ||
XM_017012092.1 | XP_016867581.1 | 10 | 1754 | 95 | ||
XM_017012093.1 | XP_016867582.1 | 10 | 1695 | 95 | ||
GenBank mRNA | AK096764.1 | - | 4 | 2434 | 95 | |
AK223406.1 | - | 12 | 2075 | 95 | ||
AK300223.1 | - | 12 | 1965 | 95 | ||
AK303819.1 | - | 10 | 1635 | 95 | ||
BC014142.2 | - | 12 | 2046 | 4662011 | 95 | |
BM719712.1 | - | 3 | 429 | 95 | ||
CD014092.1 | - | 11 | 1873 | 95 | ||
CD014093.1 | - | 10 | 1731 | 95 | ||
CD014094.1 | - | 11 | 1973 | 95 | ||
M15182.1 | - | 12 | 2071 | 95 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |