Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015445759.1 | XP_015301245.1 | 2 - 3 | 1073 | 79 | |
XM_015445760.1 | XP_015301246.1 | 3 - 4 | 480 | 79 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Probe spans exons |