Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRKAA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005545193.2 | XP_005545250.1 | 7 - 8 | 1288 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Probe spans exons |