Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LMO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005578702.2 | XP_005578759.1 | 4 - 5 | 2156 | 111 | |
XM_005578703.2 | XP_005578760.1 | 3 - 4 | 2053 | 111 | ||
XM_005578704.2 | XP_005578761.1 | 4 - 5 | 4091 | 111 | ||
XM_005578705.2 | XP_005578762.1 | 6 - 7 | 1470 | 111 | ||
XM_005578706.2 | XP_005578763.1 | 4 - 5 | 1099 | 111 | ||
XM_005578708.2 | XP_005578765.1 | 3 - 4 | 1673 | 111 | ||
XM_005578709.2 | XP_005578766.1 | 3 - 4 | 632 | 111 | ||
XM_015435174.1 | XP_015290660.1 | 4 - 5 | 1929 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Probe spans exons |