Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM53B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005566692.2 | XP_005566749.1 | 2 - 3 | 4681 | 70 | |
XM_015456829.1 | XP_015312315.1 | 3 - 4 | 1075 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Probe spans exons |