Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015449179.1 | XP_015304665.1 | 5 - 6 | 1292 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Inventoried Probe spans exons |