Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANKRD7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005550604.2 | XP_005550661.2 | 4 - 5 | 983 | 68 | |
GenBank mRNA | AB179192.1 | - | 4 - 5 | 703 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Probe spans exons |