Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ISG20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005560453.2 | XP_005560510.1 | 1 - 2 | 335 | 122 | |
XM_005560455.2 | XP_005560512.1 | 1 - 2 | 335 | 122 | ||
XM_015453365.1 | XP_015308851.1 | 1 - 2 | 335 | 122 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Probe spans exons |