Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPR19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015430395.1 | XP_015285881.1 | 1 - 2 | 879 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Probe spans exons |