Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FIZ1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005590415.2 | XP_005590472.1 | 2 - 3 | 455 | 143 | |
XM_005590416.2 | XP_005590473.1 | 2 - 3 | 1280 | 143 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Probe spans exons |