Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZFP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005592557.2 | XP_005592614.1 | 3 - 4 | 555 | 109 | |
XM_015443234.1 | XP_015298720.1 | 3 - 4 | 283 | 109 | ||
XM_015443235.1 | XP_015298721.1 | 3 - 4 | 600 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Probe spans exons |