Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GLRX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001283835.1 | NP_001270764.1 | 3 - 4 | 780 | 106 | |
XM_005540273.1 | XP_005540330.1 | 3 - 4 | 455 | 106 | ||
XM_015445324.1 | XP_015300810.1 | 4 - 5 | 460 | 106 | ||
XM_015445362.1 | XP_015300848.1 | 4 - 5 | 496 | 106 | ||
XM_015445423.1 | XP_015300909.1 | 3 - 4 | 253 | 106 | ||
GenBank mRNA | AB169463.1 | - | 3 - 4 | 780 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Probe spans exons |