Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MORF4L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001283369.1 | NP_001270298.1 | 4 | 1218 | 153 | |
XM_005594244.2 | XP_005594301.1 | 3 | 1398 | 153 | ||
XM_005594245.2 | XP_005594302.1 | 3 | 1210 | 153 | ||
XM_005594246.2 | XP_005594303.1 | 4 | 1298 | 153 | ||
XM_005594247.2 | XP_005594304.1 | 4 | 1267 | 153 | ||
XM_005594248.2 | XP_005594305.1 | 4 | 1247 | 153 | ||
XM_005594250.2 | XP_005594307.1 | 4 | 1265 | 153 | ||
XM_005594251.2 | XP_005594308.1 | 4 | 1285 | 153 | ||
GenBank mRNA | AB169286.1 | - | 1217 | 153 | ||
AB169666.1 | - | 4 | 1218 | 153 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Both primers and probe map within a single exon |