Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SYNGR3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005590923.1 | XP_005590980.1 | 1 - 2 | 169 | 76 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Probe spans exons |