Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GMEB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005544218.2 | XP_005544275.1 | 2 - 3 | 391 | 103 | |
XM_005544219.2 | XP_005544276.1 | 2 - 3 | 251 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Probe spans exons |