Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CREB3L3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015440185.1 | XP_015295671.1 | 6 - 7 | 593 | 73 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Probe spans exons |