Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYSLTR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005585841.2 | XP_005585898.2 | 5 | 2631 | 166 | |
XM_015439280.1 | XP_015294766.1 | 4 | 2532 | 166 | ||
XM_015439281.1 | XP_015294767.1 | 5 | 2028 | 166 | ||
XM_015439282.1 | XP_015294768.1 | 4 | 2509 | 166 | ||
XM_015439283.1 | XP_015294769.1 | 5 | 2608 | 166 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Inventoried Both primers and probe map within a single exon |