Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005574956.2 | XP_005575013.1 | 1 - 2 | 414 | 80 | |
XM_005574959.2 | XP_005575016.1 | 1 - 2 | 414 | 80 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Probe spans exons |