Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPRC5C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005584846.2 | XP_005584903.1 | 1 - 2 | 1675 | 121 | |
XM_015438804.1 | XP_015294290.1 | 1 - 2 | 1675 | 121 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Probe spans exons |