Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LHFPL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005550440.1 | XP_005550497.1 | 3 | 2913 | 159 | |
XM_005550441.2 | XP_005550498.1 | 4 | 2952 | 159 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Both primers and probe map within a single exon |