Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ISLR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005560050.2 | XP_005560107.1 | 2 | 2338 | 75 | |
XM_005560051.2 | XP_005560108.1 | 2 | 2146 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Both primers and probe map within a single exon |