Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADNP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005569349.1 | XP_005569406.1 | 1 - 2 | 186 | 69 | |
XM_015429972.1 | XP_015285458.1 | 1 - 2 | 1418 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Probe spans exons |