Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB11FIP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005575603.2 | XP_005575660.1 | 3 - 4 | 1729 | 140 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Probe spans exons |