Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CASS4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005569397.2 | XP_005569454.1 | 1 - 2 | 969 | 87 | |
XM_005569401.2 | XP_005569458.1 | 1 - 2 | 969 | 87 | ||
XM_005569402.2 | XP_005569459.1 | 1 - 2 | 969 | 87 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Probe spans exons |